Merck
  • Preparation of highly functionalised benzofurans from ortho-hydroxyphenones and dichloroethylene: applications and mechanistic investigations.
Preparation of highly functionalised benzofurans from ortho-hydroxyphenones and dichloroethylene: applications and mechanistic investigations.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-01-22)
Florian Schevenels, Bernard Tinant, Jean-Paul Declercq, István E Markó
ABSTRACT
Highly functionalised benzofurans have been prepared from ortho-hydroxyphenones and 1,1-dichloroethylene. The key intermediate, a chloromethylene furan, smoothly rearranged into the corresponding benzofuran carbaldehyde under acidic conditions. Some mechanistic investigations have been performed and several biologically active benzofurans have been synthesised.
Product Description
Supelco
cis-1,2-Dichloroethene, analytical standard
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%
Sigma-Aldrich
trans-1,2-Dichloroethylene, 98%
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%
Supelco
cis-1,2-Dichloroethene solution, certified reference material, 2000 μg/mL in methanol
Supelco
trans-1,2-Dichloroethylene, analytical standard