Skip to Content
Merck
All Photos(1)

Key Documents

47850

Supelco

Sodium benzoate

analytical standard

Synonym(s):

Benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E211

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

solubility

water: soluble 556 g/L

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

room temp

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium benzoate is a white granular or crystalline powdered chemical, which can be commonly be used as a food additive or preservative in food industry. It can find applications as a therapeutic agent in the treatment of some rare diseases like hyperammonemia and non-ketotic hyperglycinemia.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Davidson M.P, et al. et al.
Antimicrobials in Food (2005)
Pediatric drug formulations of sodium benzoate:: II. Coated granules with a lipophilic binder
Breitkreutz J, et al.
European Journal of Pharmaceutics and Biopharmaceutics, 56(2), 255-260 (2003)
Saurabh Khasnavis et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology, 7(2), 424-435 (2011-06-28)
DJ-1 (PARK7) is a neuroprotective protein that protects cells from oxidative stress. Accordingly, loss-of-function DJ-1 mutations have been linked with a familial form of early onset Parkinson disease. Mechanisms by which DJ-1 level could be enriched in the CNS are
Xu Huang et al.
CNS neuroscience & therapeutics, 18(1), 4-13 (2012-01-28)
D-Serine, the endogenous co-agonist of N-methyl-D-aspartate (NMDA) receptors, has been recognized as an important gliotransmitter in the mammalian brain. D-serine has been shown to prevent psychostimulant-induced decrease in hippocampal neurogenesis. However, the mechanism whereby D-serine regulates neurogenesis has not been
Sandesh C S Nagamani et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 14(5), 501-507 (2012-01-14)
The urea cycle consists of six consecutive enzymatic reactions that convert waste nitrogen into urea. Deficiencies of any of these enzymes of the cycle result in urea cycle disorders (UCDs), a group of inborn errors of hepatic metabolism that often

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service