Skip to Content
Merck
All Photos(2)

Key Documents

109169

Sigma-Aldrich

Sodium benzoate

ReagentPlus®, 99%

Synonym(s):

Benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$25.70
1 KG
$81.80
3 KG
$220.00

$25.70


Available to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$25.70
1 KG
$81.80
3 KG
$220.00

About This Item

Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39024301
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
solubility:
water: soluble(lit.)

$25.70


Available to ship on11 April 2025Details


Request a Bulk Order

grade

reagent

Quality Level

product line

ReagentPlus®

Assay

99%

mp

>300 °C (lit.)

solubility

water: soluble(lit.)

functional group

phenyl

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium benzoate is the sodium salt of benzoic acid. On dissolution in water it affords weakly basic solution. It has anti-corrosive properties.Its determination in fruit juices, sodas, soy sauce, ketchup, peanut butter, cream cheese and other foods by HPLC method has been proposed. It is a Food and Drug Administration-approved nontoxic drug. It can be obtained via acid-base reaction between benzoic acid and sodium bicarbonate/sodium hydroxide solution.

Sodium benzoate serves as a catalyst in the Knoevenagel condensation reaction to synthesize carbon carbon bond formation.[1]

Application

Sodium benzoate may be used in the preparation of various heavy metal benzoates.[2]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium benzoate as a green, efficient, and recyclable catalyst for knoevenagel condensation
Liu Q and Ai HM
Synthetic Communications, 41(20), 3004-3010 (2012)
Eagleson M.
Concise Encyclopedia Chemistry, 121-121 (1994)
Saurav Brahmachari et al.
Journal of immunology (Baltimore, Md. : 1950), 179(1), 275-283 (2007-06-21)
Experimental allergic encephalomyelitis (EAE) is the animal model for multiple sclerosis. This study explores a novel use of sodium benzoate (NaB), a commonly used food additive and a Food and Drug Administration-approved nontoxic drug for urea cycle disorders, in treating
Huey-Jen Tsay et al.
Neurotoxicology and teratology, 29(5), 562-569 (2007-07-24)
Sodium benzoate (SB) is a commonly used food preservative and anti-microbial agent in many foods from soup to cereals. However, little is known about the SB-induced toxicity and teratogenicity during early embryonic development. Here, we used zebrafish as a model
Jia Li et al.
Environmental science and pollution research international, 18(9), 1567-1573 (2011-05-18)
This study investigated the effect of copper on chemical oxygen demand (COD) removal efficiency and on the properties (mainly settling and dewatering) and the composition of extracellular polymeric substances (EPS) when 20 mg/L Cu(II) was continuously dosed to a sequencing

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service