Skip to Content
Merck
All Photos(1)

Key Documents

442658-U

Supelco

Methylene di-p-phenyl diisocyante

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
77101502

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-8°C

InChI

1S/C15H10N2O2/c18-10-16-14-5-1-12(2-6-14)9-13-3-7-15(8-4-13)17-11-19/h1-8H,9H2

InChI key

UPMLOUAZCHDJJD-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

411.8 °F - closed cup

Flash Point(C)

211 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanna K Lindberg et al.
Mutation research, 723(1), 1-10 (2011-04-02)
Toluene diisocyanate (TDI) and 4,4'-methylenediphenyl diisocyanate (MDI), used in the production of polyurethane foam, are well known for their irritating and sensitizing properties. Contradictory results have been obtained on their genotoxicity. We investigated the genotoxicity and protein binding of inhaled
Dominika Swierczyńska-Machura et al.
Medycyna pracy, 62(6), 567-577 (2012-02-09)
The aim of the study was to analyze the data obtained during the diagnostic process and their real values in the final diagnosis of patients exposed to diisocyanates amd suspected of occupational allergic disease of the respiratory system The patients
Unique foreign body ingestions in the pediatric population.
Sheila Isabelle Goertemoeller et al.
Journal of emergency nursing: JEN : official publication of the Emergency Department Nurses Association, 36(2), 178-179 (2010-03-10)
H D Hoffmann et al.
Toxicology letters, 199(3), 364-371 (2010-10-12)
Polyurethanes (PU) are polymers made with diisocyanates such as MDI (4,4'-methylene diphenyl diisocyanate) and TDI (2,4-toluene diisocyanate and 2,6-toluene diisocyanate). Investigations have been undertaken with MDI and TDI to assess dermal uptake and resulting systemic exposure. Absorption, distribution and excretion
Gabriele Sabbioni et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 15(6), 508-515 (2010-06-18)
4,4'-Methylenediphenyl diisocyanate (MDI) is the most important of the isocyanates used as intermediates in the chemical industry. Among the main types of damage after exposure to low levels of MDI are lung sensitization and asthma. Albumin adducts of MDI might

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service