Skip to Content
Merck
All Photos(1)

Key Documents

17969

Sigma-Aldrich

Diethylenetriamine-pentaacetic acid pentasodium salt solution

purum, ~40% in H2O

Synonym(s):

DTPA Na5

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$77.10

$77.10


In StockDetails


Request a Bulk Order

Select a Size

Change View
500 ML
$77.10

About This Item

Linear Formula:
[(NaOOCCH2)2NCH2CH2]2NCH2COONa · aq
CAS Number:
Molecular Weight:
503.26 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

$77.10


In StockDetails


Request a Bulk Order

grade

purum

Quality Level

concentration

~40% in H2O

functional group

amine

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O

InChI

1S/C14H23N3O10.5Na/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27;;;;;/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27);;;;;/q;5*+1/p-5

InChI key

LQPLDXQVILYOOL-UHFFFAOYSA-I

General description

Diethylenetriamine-pentaacetic acid pentasodium salt is an organic chelating agent.

Application

Diethylenetriamine-pentaacetic acid pentasodium salt solution may be used in a competitive indirect enzyme-linked immunoassay for the quantitative measurement of lead ions.[1] It may be used in one-step immunochromatographic assay (ICA) for the fast detection of lead ions in water samples.[2]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Repr. 1B - STOT RE 2 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Tang et al.
Environmental pollution (Barking, Essex : 1987), 158(6), 2074-2077 (2010-04-07)
One-step immunochromatographic assay (ICA) has been developed using colloidal gold-labeled monoclonal antibody probe for the rapid detection of lead ions in water samples. The ICA was based on the theory of competitive reactivity, and the results can be easily judged
Jun-jian Xiang et al.
Environmental pollution (Barking, Essex : 1987), 158(5), 1376-1380 (2010-02-09)
Immunoassays for quantitative measurement of environmental heavy metals offer several advantages over other traditional methods. To develop an immunoassay for lead, Balb/c mice were immunized with a lead-chelate-protein conjugate to allow maximum exposure of the metal to the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service