Skip to Content
Merck
All Photos(1)

Key Documents

29441-U

Supelco

Certified glass inserts for 12 x 32 mm, large opening vials

volume 0.35 mL, clear glass insert (flat bottom), O.D. × H 6 mm × 31 mm, pkg of 100 ea

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$27.60

$27.60


Available to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 EA
$27.60

About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

$27.60


Available to ship on24 April 2025Details


Request a Bulk Order

material

clear glass insert (flat bottom)

description

flat bottom

packaging

pkg of 100 ea

O.D. × H

6 mm × 31 mm

volume

0.35 mL

fitting

insert for large opening vials (6.0 mm I.D.)

Looking for similar products? Visit Product Comparison Guide

Other Notes

Large opening inserts fits crimp seal, snap ring, and 9 mm threaded vials (with 6.0 mm ID)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Stravs et al.
Environmental science & technology, 53(8), 4265-4273 (2019-03-16)
Biotransformation of chemical pollutants is an ecological process requiring multifunctionality (multiple metabolic pathways) and, potentially, high biodiversity. Phytoplankton communities are highly diverse functionally and taxonomically and co-occur with complex mixtures of organic pollutants in aquatic environments. Here, we investigated how

Articles

HPLC method determines Drospirenone in progestin medication using Purospher® STAR RP-18e Hibar® HPLC column, meeting USP monograph criteria.

Learn how the Purospher® STAR RP-18e Hibar® HPLC column meets the system suitability criteria outlined in the USP monograph for the fluticasone propionate assay.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service