Skip to Content
Merck
All Photos(1)

Key Documents

28987-U

Supelco

Purge/Trap H

configured for Tekmar® Stratum

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$225.00

$225.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$225.00

About This Item

UNSPSC Code:
12000000
NACRES:
NB.21

$225.00


Check Cart for Availability

Request a Bulk Order

application(s)

industrial hygiene

compatibility

configured for Tekmar® Stratum

Looking for similar products? Visit Product Comparison Guide

Legal Information

Tekmar is a registered trademark of Tekmar Co.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Are the Supelco Purge Traps for Tekmar Stratum compatible with the Tekmar Atomx?

    1 answer
    1. Purge traps manufactured for the Tekmar Stratum PTC Purge and Trap Concentrator are the same traps used for the Tekmar Atomx Automated Sample Prep System.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service