Skip to Content
Merck
All Photos(2)

Key Documents

535494

Sigma-Aldrich

4′-Bromomethyl-2-biphenylcarbonitrile

97%

Synonym(s):

2-Cyano-4′-bromomethylbiphenyl, 4′-(Bromomethyl)-2-cyanobiphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$131.00
25 G
$436.00

$131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$131.00
25 G
$436.00

About This Item

Linear Formula:
BrCH2C6H4C6H4CN
CAS Number:
Molecular Weight:
272.14
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$131.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid crystal

mp

125-128 °C (lit.)

SMILES string

BrCc1ccc(cc1)-c2ccccc2C#N

InChI

1S/C14H10BrN/c15-9-11-5-7-12(8-6-11)14-4-2-1-3-13(14)10-16/h1-8H,9H2

InChI key

LFFIEVAMVPCZNA-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service