Skip to Content
Merck
All Photos(1)

Key Documents

28522-U

Supelco

ATIS Purge and Trap/Humidifier Module

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,390.00

$1,390.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$1,390.00

About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

$1,390.00


Check Cart for Availability

Request a Bulk Order

application(s)

air monitoring
environmental
industrial hygiene

General description

A separate module is available that will allow you to purge aqueous samples onto an adsorbent tube at ambient temperatures. This module can also be used to generate a dynamic humidified stream of the carrier gas for spiking calibration standards. The purge and trap module includes purge and trap glassware, and a separate flow controller that allows the user to set a separate purge (wet) flow rate independently of the dry flow rate. The purge and trap module accepts standard 22mL threaded vials to simplify your sample prep.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service