Skip to Content
Merck
All Photos(1)

Key Documents

2632705

Supelco

Inlet Liner, Split/Splitless Type, Straight Design (wool packed)

pkg of 5 ea

Sign Into View Organizational & Contract Pricing

Select a Size

5 PKG
$227.00

$227.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 PKG
$227.00

About This Item

UNSPSC Code:
41115719

$227.00


Check Cart for Availability

Request a Bulk Order

description

packed (deactivated glass wool)

packaging

pkg of 5 ea

L × O.D. × I.D.

95 mm × 5.0 mm × 3.0 mm

compatibility

technique used for Dirty samples

Looking for similar products? Visit Product Comparison Guide

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Shimadzu (17A) [with SPL-17 Injector]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service