21313
Hamilton® Microliter™ syringe, cemented needle
701 ASN, volume 10 μL, needle size 23s ga (cone tip), needle L 43 mm (1.71 in.), pkg of 1 ea
Select a Size
About This Item
Recommended Products
material
cone tip stainless steel needle (point style 1)
needle size
23s ga (cone tip)
feature
needle type N (cemented needle)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80387
needle L
43 mm (1.71 in.)
volume
10 μL
compatibility
for use with Agilent/HP 7673 and 7683 autosampler
Related Categories
General description
The plunger is not to be used with any other syringe.
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)