25339
Intermediate Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.53 mm
Select a Size
$184.00
Select a Size
About This Item
$184.00
Recommended Products
material
fused silica tubing
Quality Level
description
Intermediate Polarity Fused Silica Tubing, phenyl/methyl deactivated
composition
phenyl/methyl (deactivated)
packaging
pkg of 1 ea
technique(s)
gas chromatography (GC): suitable
L × I.D.
5.0 m × 0.53 mm
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Static headspace GC (GC-SH) concentrates volatile analytes for improved detection and minimizes matrix interference.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service