Key Documents
23627
Supelco
GlasSeal Capillary Column Connector, Fused Silica
pkg of 5 ea
Select a Size
5 EA
$159.00
$159.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
5 EA
$159.00
About This Item
UNSPSC Code:
41115700
$159.00
Check Cart for Availability
Request a Bulk Order
packaging
pkg of 5 ea
technique(s)
gas chromatography (GC): suitable
General description
Quickly connect two pieces of fused silica tubing of the same or different diameters without the need for tools. The leak-free connection is useful to connect a guard column or transfer line, repair a broken column, or connect two columns (same or different phases). Silanized for an inert inside surface. For use with 0.10 - 0.53 mm I.D. fused silica tubing.
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
R R Heath et al.
Journal of chemical ecology, 19(10), 2395-2410 (1993-10-01)
A bioassay system was developed that permits the testing of various substrates for biological activity in a flight tunnel, while simultaneously collecting a portion of the volatiles from the attractive source for subsequent chemical identification and quantification. Bioassays of the
A T Fisk et al.
Archives of environmental contamination and toxicology, 42(1), 118-126 (2001-11-14)
To examine the influence of diet and age on organochlorine contaminant (OC) concentrations in two closely related ringed seal (Phoca hispida) populations enantiomeric fractions (EFs) of chiral contaminants and stable isotopes of nitrogen (delta15N) and carbon (delta13C) were measured along