Skip to Content
Merck
All Photos(1)

Key Documents

22688

Swagelok® Port Connector

Swagelok® 201-PC, brass, 1/8 in. Swagelok

Synonym(s):

Swagelok® 201-PC,BRASS PORT CONNECTOR 1/8" PK2

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$42.00

$42.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 EA
$42.00

About This Item

UNSPSC Code:
41115717

$42.00


Check Cart for Availability

Request a Bulk Order

material

brass

packaging

pkg of 2 ea

manufacturer/tradename

Swagelok® 201-PC

fitting

1/8 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

Application

Swagelok® port connector with two Swagelok® end caps may be used in construction of the reactors used in experimental studies investigating alkylarene pyrolysis coupled with mechanistic modeling.[1]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of polycyclic alkylaromatics. 4. Hydrogenolysis mechanisms in 1-alkylpyrene pyrolysis
Smith, C. Michael, and Phillip E. Savage
Energy and Fuels, 6, 195-202 (1992)
Cody J Warren et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(23), 11460-11469 (2019-05-23)
Pandemic HIV-1 (group M) emerged following the cross-species transmission of a simian immunodeficiency virus from chimpanzees (SIVcpz) to humans. Primate lentiviruses (HIV/SIV) require the T cell receptor CD4 to enter into target cells. By surveying the sequence and function of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service