Skip to Content
Merck
All Photos(1)

Key Documents

22554

Supelco

Vinyl chloride in nitrogen

1 ppm, cylinder of 14 L, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

14 L
$403.00

$403.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
14 L
$403.00

About This Item

UNSPSC Code:
41116107

$403.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

form

gas

packaging

cylinder of 14 L

manufacturer/tradename

Scott Gas T70-14

availability

not available in Canada, EU

concentration

1 ppm

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

  • Vinyl chloride in nitrogen, 1 ppm is a two component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.

Pictograms

Gas cylinderHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service