Key Documents
20352
Septum Puller
hook design
Select a Size
1 EA
$183.00
$183.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$183.00
About This Item
UNSPSC Code:
47131900
$183.00
Check Cart for Availability
Request a Bulk Order
packaging
pkg of 1 ea
technique(s)
gas chromatography (GC): suitable
L
4 in.
General description
The hook septum puller is great for removing soft silicone septa from injection ports, and graphite ferrules from columns nuts. It has dozens of other uses around the lab.
Choose from one of the most recent versions:
Certificates of Analysis (COA)