Key Documents
22495-U
Supelco
Supeltex M-4 Packed Column Ferrule, 1/16 in. Column O.D.
I.D. 1/16 in., 1/16 in. O.D. column, pkg of 10 ea
Select a Size
10 EA
$117.00
$117.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
10 EA
$117.00
About This Item
UNSPSC Code:
41121815
NACRES:
SB.54
$117.00
Check Cart for Availability
Request a Bulk Order
packaging
pkg of 10 ea
maximum temperature
450 °C
I.D.
1/16 in.
compatibility
1/16 in. O.D. column
General description
Ferrules are suitable for the attachment of the column and other glass components to the system. Flexible graphite ferrules withstand high temperatures and are capable of compression.[1]
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics:
Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Walter Jennings
Gas Chromatography with Glass Capillary Columns (2012)