Skip to Content
Merck
All Photos(1)

Documents

20425-U

Sigma-Aldrich

Heavy Duty Tubing Cutter

manual, tubing cutter L 4 15/16 in., pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122406
NACRES:
NB.35

feature

manual

packaging

pkg of 1 ea

tubing cutter L

4 15/16 in.

Features and Benefits

Cuts metal tubing 1/8in. to 1-1/8in. O.D.
Spare cutting wheel included.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service