Key Documents
22069
Swagelok® PTFE Ferrules
PTFE back ferrule, Swagelok for 1/16 in. tubing
Select a Size
5 EA
$58.60
$58.60
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
5 EA
$58.60
About This Item
UNSPSC Code:
41115717
$58.60
Check Cart for Availability
Request a Bulk Order
material
PTFE back ferrule
packaging
pkg of 5 ea
fitting
Swagelok for 1/16 in. tubing
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1]
Use PTFE ferrules instead of brass or stainless steel ferrules when attaching fittings to glass tubing, or to flexible tubing (such as PTFE or Tygon).
Application
Swagelok® PTFE ferrules maybe used, for making all the connections between glass and PTFE present in the containment vessels, piping and sampling equipment in a bench scale biofilter system, in an experimental study investigating the effects of periods of non-use on biofilter performance.[2]
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Effect of periods of non-use on biofilter performance.
Jason MF and Loehr RC.
Journal of the Air & Waste Management Association (1995), 46 (6), 539-546 (1996)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)