Skip to Content
Merck
All Photos(1)

Key Documents

20012-U

Supelco

Carbosieve SIII

stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

material

stainless steel TD tube

Agency

EPA TO-2

description

Sealed with TDS³ Storage Container

product line

Carbosieve

feature

preconditioned

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Carbosieve S-III carbon molecular sieve

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

Looking for similar products? Visit Product Comparison Guide

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yannick Juillet et al.
The Analyst, 130(6), 977-982 (2005-05-25)
Among the chemicals belonging to the schedules of the Chemical Weapons Convention (CWC), sampling and analysis of highly volatile compounds such as hydrogen cyanide (HCN) require special consideration. The latter is present in numerous old chemical weapons that are stockpiled
Juliane Hollender et al.
Journal of chromatography. A, 962(1-2), 175-181 (2002-08-30)
A simple method using active trapping on adsorbents and thermal desorption followed by GC-MS analysis was developed for the indoor air monitoring of monoterpenes. The study was carried out using a dynamically generated atmosphere consisting of 11 monoterpenes: camphene, camphor
C Y Peng et al.
Journal of environmental monitoring : JEM, 2(4), 313-324 (2001-03-16)
Air sampling, using sorbents, thermal desorption and gas chromatography, is a versatile method for identifying and quantifying trace levels of volatile organic compounds (VOCs). Thermal desorption can provide high sensitivity, appropropriate choices of sorbents and method parameters can accommodate a
John L Martin et al.
Journal of occupational and environmental hygiene, 1(6), 363-370 (2004-07-09)
In an effort to assess the scope of occupational exposures to ethylene, the Olefins Panel of the American Chemistry Council designed and conducted a research project to develop and apply a sampling and analytical method to measure workplace exposure. The
Mohammad Reza Samarghandi et al.
Journal of research in health sciences, 14(3), 227-232 (2014-09-12)
Toluene is a volatile organic compound, one of 189 hazardous air pollutants (HAPs) and the most important pollutant found in most industries and indoor environments; owing to its adverse health, toluene must be treated before being released into the environment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service