The shelf life of the Orbo-91 active air monitoring tube is two years.
20360
ORBO™ 91 Carbosieve-SIII specially treated (60/80), 130/65 mg
W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea
About This Item
Recommended Products
material
W,W,W separators
Agency
OSHA 69,75,80,84,88,91,100,103,106,111,113
product line
ORBO™
composition
Bed A, 130 mg
Bed B, 65 mg
packaging
pkg of 25 ea
manufacturer/tradename
ORBO™ 91
technique(s)
active air sampling: suitable
O.D. × L
6 mm × 75 mm
matrix
Carbosieve S-III Carbon Molecular Sieve (CMS) (specially treated)
particle size
20-45 mesh
application(s)
air monitoring
environmental
industrial hygiene
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.
-
What is the expiration date for the product Orbo-91, an air monitoring tube used in several OSHA methods?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service