Skip to Content
Merck
All Photos(1)

Key Documents

17811-1AMP

Supelco

Lipid standards: triglyceride mixtures

certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$141.00

$141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$141.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

$141.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

functional group

ester

shipped in

dry ice

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Glyceryl tridecanoate ~ 20 mg
    Glyceryl tridodecanoate ~ 20 mg
    Glyceryl trimyristate ~ 20 mg
    Glyceryl trioctanoate ~ 20 mg
    Tripalmitin ~ 20 mg

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenqiang Chang et al.
Scientific reports, 5, 15133-15133 (2015-10-16)
Lipid droplets (LDs) act as intracellular storage organelles in most types of cells and are principally involved in energy homeostasis and lipid metabolism. However, the role of LDs in resistance to toxins in fungi remains largely unknown. Here, we show
Fatima Ameer et al.
Biochimie, 135, 104-110 (2017-02-06)
Lipid-load in peripheral blood mononuclear cells (PBMCs) has recently gained attention of the researchers working on nutritional regulation of metabolic health. Previous works have indicated that the metabolic circuitries in the circulating PBMCs are influenced by dietary-intake and macronutrient composition
Satoshi Yoshida et al.
Lipids in health and disease, 8, 28-28 (2009-07-25)
The metabolism of dietary fatty acids in human has been measured so far using human blood cells and stable-isotope labeled fatty acids, however, no direct data was available for human peripheral tissues and other major organs. To realize the role
Jan Lisec et al.
BMC cancer, 19(1), 501-501 (2019-05-30)
Cancer cells modify the balance between fatty acid (FA) synthesis and uptake under metabolic stress, induced by oxygen/nutrient deprivation. These modifications were shown to alter the levels of individual triglyceride (TG) or phospholipid sub-species. To attain a holistic overview of
William M Dickerson et al.
PloS one, 16(8), e0256423-e0256423 (2021-08-27)
Point-of-care (PoC) testing of platelet count (PLT) provides real-time data for rapid decision making. The goal of this study is to evaluate the accuracy and precision of platelet counting using a new microvolume (8 μL), absolute counting, 1.5 kg cytometry-based

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service