Skip to Content
Merck
All Photos(1)

Key Documents

O7631-1AMP

Supelco

F.A.M.E. Mix RM-6

certified reference material, pkg of 100 mg (Neat)

Synonym(s):

AOCS Reference Mixtures, AOCS No. 6 (RM-6), Oil Reference Standard, AOCS No. 6, Oil Reference Standard, AOCS

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
$123.00

$123.00


Available to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 AMP
$123.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

$123.00


Available to ship on28 April 2025Details


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 100 mg fatty acid methyl ester mixture
pkg of 100 mg (Neat)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

carboxylic acid
ester

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Our quantitative animal and vegetable reference mixes conform to the requirements of the American Oil Chemists′ Society (AOCS) method Ce 1-62. The composition of each mix is similar to the fatty acid distribution of certain oils listed under each product. Please note that the AOCS mixtures sold under the Supelco brand are named RM-1, RM-2, etc.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methyl linoleate 7 wt. %
    Methyl linolenate 3 wt. %
    Methyl myristate 2 wt. %
    Methyl oleate 41 wt. %
    Methyl palmitate 30 wt. %
    Methyl palmitoleate 3 wt. %
    Methyl stearate 14 wt. %

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service