Skip to Content
Merck
All Photos(1)

Key Documents

1505-U

Supelco

Coated GC Packing

phase 1.5% OV-225 + 1% H3PO4, matrix 60/80 Carbopack B support, bottle of 10 g

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$694.00

$694.00


Check Cart for Availability


Select a Size

Change View
10 G
$694.00

About This Item

EC Number:
UNSPSC Code:
23151817
NACRES:
SB.54

$694.00


Check Cart for Availability

packaging

bottle of 10 g

parameter

60-220 °C temp. limit

technique(s)

gas chromatography (GC): suitable

matrix

60/80 Carbopack B support

matrix active group

1.5% OV-225 + 1% H3PO4 phase

column type

packed GC

compatibility

for analyte group amines

Looking for similar products? Visit Product Comparison Guide

General description

Ready-to-use coated packing for those wanting to pack their own columns.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service