Skip to Content
Merck
All Photos(1)

Key Documents

SML0397

Sigma-Aldrich

DR2313

≥98% (HPLC)

Synonym(s):

3,5,7,8-Tetrahydro-2-methyl-4H-thiopyrano[4,3-d]pyrimidin-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$67.30

$67.30


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$67.30

About This Item

Empirical Formula (Hill Notation):
C8H10N2OS
CAS Number:
Molecular Weight:
182.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$67.30


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: >2 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

CC1=NC(=O)C2=C(CCSC2)N1

InChI

1S/C8H10N2OS/c1-5-9-7-2-3-12-4-6(7)8(11)10-5/h2-4H2,1H3,(H,9,10,11)

InChI key

HRYKZAKEAVZGJD-UHFFFAOYSA-N

Biochem/physiol Actions

DR2313 is a PARP inhibitor.
DR2313 is a PARP-1; PARP-2 inhibitor (Ki = 0.23 mM). DR2313 has neuroprotective effects in both in vivo and in vitro ischemic models, and reduces cortical infarct volume in focal ischemia in rats.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luyun Zou et al.
Frontiers in molecular biosciences, 8, 780865-780865 (2021-12-25)
The modification of proteins by O-linked β-N-acetylglucosamine (O-GlcNAc) is associated with the regulation of numerous cellular processes. Despite the importance of O-GlcNAc in mediating cellular function our understanding of the mechanisms that regulate O-GlcNAc levels is limited. One factor known

Articles

Agents reported to activate cellular caspases include chemotherapeutic drugs, TNF receptor agonists, and other enzymes. Inhibitors of apoptosis were the first identified endogenous caspase inhibitors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service