Skip to Content
Merck
All Photos(1)

Key Documents

13349-U

Supelco

Dowex Monosphere MR-450 UPW hydrogen and hydroxide form

matrix styrene-divinylbenzene (gel)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

form

beads

Quality Level

packaging

bucket of 1000 g

parameter

60 °C max. temp.

moisture

46-53% (cation)
55-65% (anion)

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

particle size

310-410 μm (cation exchange)
540-640 μm (anion exchange)

operating pH

0-14

capacity

1.0 meq/mL by wetted bed volume(anion)
1.9 meq/mL by wetted bed volume(cation)

separation technique

mixed mode

General description

Dowex Monosphere MR-450 is a gel-type nonseparable bifuntional styrenedivinylbenzene mixed bed ion exchange resin. It has sulfonic acid and quaternary ammonium as functional groups. It is used for adsorptive deacidification of fatty acid oils.[1]

Application

Dowex Monosphere MR-450 UPW hydrogen and hydroxide form may be used in separation media for liquid chromatography.
Mixed bed ion exchange resin capable of deionizing for ultrapure water.

Legal Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sorption Kinetics, Thermodynamics and Regeneration for Lipid Feedstock Deacidification Using a Mixed-Bed Ion-Exchange Resin
Jamal, Yousuf, et al.
Journal of Food Process Engineering, 37, 27-36 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service