Skip to Content
Merck
All Photos(1)

Key Documents

01387

Supelco

Agaric acid

analytical standard

Synonym(s):

2-Hydroxy-1,2,3-nona­decane­tricarboxylic acid, Agaricic acid, Agaricin

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$165.00

$165.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$165.00

About This Item

Empirical Formula (Hill Notation):
C22H40O7
CAS Number:
Molecular Weight:
416.55
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$165.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

impurities

≤8.0% water (calculated from elemental analysis)

SMILES string

CCCCCCCCCCCCCCCCC(C(O)=O)C(O)(CC(O)=O)C(O)=O
CCCCCCCCCCCCCCCCC(C(O)=O)C(O)(CC(O)=O)C(O)=O

InChI

1S/C22H40O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-18(20(25)26)22(29,21(27)28)17-19(23)24/h18,29H,2-17H2,1H3,(H,23,24)(H,25,26)(H,27,28)

InChI key

HZLCGUXUOFWCCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service