Skip to Content
Merck
All Photos(13)

Key Documents

ZRB2029

Sigma-Aldrich

Anti-MRP2 Antibody, clone 1B2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ATP-binding cassette sub-family C member 2;Canalicular multidrug resistance protein;Canalicular multispecific organic anion transporter 1;Multidrug resistance-associated protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$357.00
4 X 25 μL
$735.00

$357.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$357.00
4 X 25 μL
$735.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$357.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B2, recombinant monoclonal

description

1B2 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 174.21 kDa
observed mol wt ~200 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ABCC2(1244)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B2 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Multidrug resistance-associated protein 2 ( MRP2). It targets an epitope within 14 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal, cytoplasmic domain of human Multidrug resistance-associated protein 2 ( MRP2).

Application

Quality Control Testing

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MRP2 in A549 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MRP2 in Human liver tissue lysate.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected MRP2 in one million A549 cells.

Affinity Binding Assay: A representative lot of this antibody bound MRP2 peptide with a KD of 1.3 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MRP2 in A549 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:500 dilution from a representative lot detected MRP2 in human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

ATP-binding cassette sub-family C member 2 (UniProt: Q92887; also known as Canalicular multidrug resistance protein, Canalicular multi-specific organic anion transporter 1, Multidrug resistance-associated protein 2, MRP2) is encoded by the ABCC2 (also known as CMOAT, CMOAT1, CMRP, MRP2) gene (Gene ID: 1244) in human. MRP2, a member of the family of human ATP-binding cassette (ABC) transporters, binds and hydrolyzes ATP to enable active transport of various substrates, including many drugs, toxicants, and endogenous compound across cell membranes. It transports a wide variety of conjugated organic anions such as sulfate-, glucuronide- and glutathione (GSH)-conjugates of endo- and xenobiotics substrates. It is reported to mediate hepatobiliary excretion of mono- and bis-glucuronidated bilirubin molecules. Hence, it plays an important role in bilirubin detoxification. MRP2 is expressed in the apical hepatocyte plasma membrane, renal proximal tubules, and small intestine, where it plays a role in the elimination and oral bioavailability of drugs, xenotoxins, and their metabolites. It also confers resistance to several anti-cancer drugs, including cisplatin, doxorubicin, epirubicin, methotrexate, etoposide, and vincristine. It contains two conserved nucleotide-binding domains (aa 671-678 and 1334-1341) that mediate the binding and hydrolyzing ATP via an ATPase, thereby providing energy for substrates export from the cytoplasm to the extracellular space. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hayashi, H., et al. (2005). Biochim. Biophys. Acta. 1738(1-3); 54-62; Cui, Y., et al. (1999). Mol. Pharmacol. 55(5); 929-937; Kamisako, T., et al. (1999). Hepatology. 30(2); 485-490).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service