Skip to Content
Merck
All Photos(10)

Key Documents

ZRB06182

Sigma-Aldrich

Anti-p44 MAPK (ERK1) Antibody, clone 13F10, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC: 2.7.11.24, ERK-1, ERT2, Extracellular signal-regulated kinase 1, Insulin-stimulated MAP2 kinase, MAP kinase 3, MAP kinase isoform p44, MAPK 3, MNK1, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 3, p44-ERK1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$303.00
4 X 25 μL
$625.00

$303.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
$303.00
4 X 25 μL
$625.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$303.00


Available to ship on22 April 2025Details


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

13F10, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 43.08 kDa
observed mol wt ~44 kDa

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using 1:100
immunoprecipitation (IP): suitable using 1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 13F10 is a ZooMAb rabbit recombinant monoclonal antibody that detects p44 MAPK (ERK1). May also display reactivity with p42 MAPK (ERK2).

Immunogen

KLH-conjugated linear peptide corresponding to 35 amino acids from the C-terminal region of rat p44 MAPK (ERK1).

Application

Anti-p44 MAPK (ERK1), clone 13F10 ZooMAb, Cat. No. ZRB06182, is a recombinant Rabbit Monoclonal Antibody that detects p44 MAPK (ERK1) and has been tested for use in Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Western Blotting Analysis: 1:1,000 dilution from a representative lot detected p44 MAPK (ERK1) in Rat C6, L6, and mouse NIH/3T3 cell lysates.

Immunoprecipitation Analysis: 100 μL from a representative lot immunoprecipitated p44 MAPK (ERK1) in Rat L6 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected p44 MAPK (ERK1) in rat colon and rat cerebral cortex tissue sections.

Target description

Mitogen-activated protein kinase 3 (UniProt: P21708; also known as EC: 2.7.11.24, MAP kinase 3, MAPK 3, ERT2, Extracellular signal-regulated kinase 1, ERK-1, Insulin-stimulated MAP2 kinase, MAP kinase isoform p44, p44-MAPK, MNK1, Microtubule-associated protein 2 kinase, p44-ERK1) is encoded by the Mapk3 (also known as Erk1, Prkm3) gene (Gene ID: 50689) in rat. ERK-1 is a serine/threonine kinase that acts as an essential component of the MAP kinase signal transduction pathway. It is expressed in different tissues, mostly in intestine, placenta, and lung. Highest levels have been reported in the nervous system. The MAPK/ERK cascade mediates several diverse biological processes such as cell growth, adhesion, survival, and differentiation through the regulation of transcription, translation, and cytoskeletal rearrangements. It also plays also a role in initiation and regulation of meiosis, mitosis, and post-mitotic functions in differentiated cells by phosphorylating a few transcription factors. About 160 substrates have been described for ERKs. The protein kinase domain of ERK-1 is localized to amino acids 43-331 and the TXY motif is located in amino acids 203-205. ERK-1 is phosphorylated by MEK1 and MEK2 on threonine 203 and tyrosine 205 in response to external stimuli like insulin or nerve growth factor and both these phosphorylations are essential for its activity. Two isoforms of ERK-1 have been described that are produced by alternative splicing. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service