ZRB1888
Anti-Histone H3-Lys9me3 Antibody, clone 4J8 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Histone H31t;H3/t;H3/g;H3-Lys9 3me, H3K9me3
Select a Size
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
4J8, recombinant monoclonal
description
4J8 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 15.51 kDa
observed mol wt ~16 kDa
purified by
using Protein A
species reactivity
mouse, rat, human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
trimethylation (Lys9)
Gene Information
human ... HIST3H3(8290)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in HeLa cell acid extract.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Histone H3-Lys9me3 in HeLa cell acid extract.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Histone H3-Lys9me3 in Balb3T3/A31 and RAT2 cell lysates.
Affinity Binding Assay: A representative lot of this antibody bound Tri-Methyl-Histone H3-Lys9 peptide with a KD of 1.9 x 10-6 in an affinity binding assay.
Peptide Inhibition Assay: Target band detection in HeLa cell acid extract was prevented by pre-blocking of a representative lot with Histone H3-Lys9me3 peptide, but not with non-methylated Histone H3-Lys9
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Histone H3-Lys9me3 in human colon tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service