Key Documents
Z763462
MAGic Clamp universal platform for flasks and tube racks
L × W 18 in. × 18 in.
Synonym(s):
magnetic clamps, orbital shaker platforms
Select a Size
1 EA
$608.00
$608.00
Check Cart for Availability
Select a Size
Change View
1 EA
$608.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.77
$608.00
Check Cart for Availability
manufacturer/tradename
Benchmark Scientific H1010-MR
L × W
18 in. × 18 in.
General description
The convenient MAGic Clamp platforms (large or small) are available for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is widely recognized as the easiest way to instantly change between flask clamps of different sizes.
Legal Information
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)