Documents
Z763403
Orbi-Shaker XL orbital shaker with flat mat platform
AC/DC input 230 V AC, UK plug
Synonym(s):
orbital mixers, orbital shakers, stirring plates
About This Item
UNSPSC Code:
41103811
NACRES:
NB.77
manufacturer/tradename
Benchmark Scientific BT1011-E-UK
parameter
30-300 rpm speed
overall L × W × H
18.5 in. (47 cm) × 21.25 in. (54 cm) × 5 in. (12.5 cm)
platform L × W
17.8 in. (45 cm) × 17.8 in. (45 cm)
weight
30 kg
General description
The Orbi-Shaker XL offers flask capacity that far exceeds most bench-top models. With the large platform and powerful, beltless motor drive, the Orbi-Shaker XL is capable of mixing up to 5 x 2 L flasks or 9 x 1 L flasks simultaneously. The included non-slip rubber mat platform is ideal for low speed applications, while the convenient MAGic Clamp platform is also available for high speed mixing of Erlenmeyer flasks, tube racks or microplates. The MAGic Clamp allows for instant flask clamp exchange without the use of tools, screws or any other hardware.
Legal Information
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
Orbi-Shaker is a trademark of Benchmark Scientific, Inc.