Key Documents
Z761036
TPP® TubeSpin bioreactor bottles
capacity 600 mL
Synonym(s):
Bioreactor for cell culture, TPP bioreactor, TubeSpin bioreactor
Select a Size
26 EA
$1,720.00
$1,720.00
Available to ship on14 April 2025Details
Select a Size
Change View
26 EA
$1,720.00
About This Item
UNSPSC Code:
41122105
NACRES:
NB.22
$1,720.00
Available to ship on14 April 2025Details
packaging
pack of 26 ea
manufacturer/tradename
TPP 87600
capacity
600 mL
size
General description
TPP® TubeSpin bioreactor bottles are designed for large-scale screening, optimization, and process of cell suspension system. These bioreactor bottles offer several advantages, including the ability to evaluate small amounts of important parameters during production upscaling, thereby maximizing the number of parallel experiments at the same time. This approach effectively lowers cost and increases the efficiency of the process.
Features and Benefits
  • Conical bottom with filter cap
  • Fit standard centrifuge apparatus
Legal Information
TPP is a registered trademark of TPP Techno Plastic Products AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)