Skip to Content
Merck
All Photos(1)

Key Documents

B1408

Nalgene® centrifuge bottles, style 3120

nominal capacity 1000 mL

Synonym(s):

Bioprocessing Containers, purification of large cell volumes

Sign Into View Organizational & Contract Pricing

Select a Size

4 EA
$364.00

$364.00


Available to ship on11 April 2025Details



Select a Size

Change View
4 EA
$364.00

About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

$364.00


Available to ship on11 April 2025Details


material

polypropylene copolymer bottle
round bottle
screw top cap
translucent bottle

Joint

Joints threaded neck (52.6)

feature

RCF 7,100 × g
with cap

packaging

pack of 4

manufacturer/tradename

Nalgene® 3120-1000

parameter

7100 × g max. RCF

bottle diam. × L

98.04 mm × 174.2 mm

nominal capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Perform biopharmaceutical aseptic fluid transfer or sampling applications with the tri-clover flange of PPCO Centrifuge Bottles, molded of translucent polypropylene copolymer. Ideal for large-volume cell harvesting, pelleting and protein purification, these translucent bottles are autoclavable and have excellent chemical resistance properties.

Application

Nalgene® centrifuge bottles, style 3120 are ideal for the transfer of biopharmaceutical aseptic fluid and sampling applications. They are also suitable for large-volume cell harvesting, protein purification, and pelleting.

Features and Benefits

  • Autoclave friendly and chemically resistant
  • Ideal for low and moderate-speed centrifugation
  • Leakproof at higher speed and volume
  • 80% of the total fill capacity enables better performance

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra M Welker et al.
Disease models & mechanisms, 9(2), 199-210 (2015-12-15)
Glioblastoma (GBM) is a deadly brain cancer, for which few effective drug treatments are available. Several studies have used zebrafish models to study GBM, but a standardized approach to modeling GBM in zebrafish was lacking to date, preventing comparison of
Hotaka Kobayashi et al.
Cell reports, 28(5), 1144-1153 (2019-08-01)
The Argonaute subfamily of proteins (AGO) loads microRNAs (miRNAs) to form the effector complex that mediates target gene silencing. Empty AGO, but not miRNA-loaded AGO, is selectively degraded across species. We have reported that the degradation of empty AGO is
Margarita Cabrera et al.
Cell reports, 30(7), 2430-2443 (2020-02-23)
Cells have developed protein quality-control strategies to manage the accumulation of misfolded substrates during heat stress. Using a soluble reporter of misfolding in fission yeast, Rho1.C17R-GFP, we demonstrate that upon mild heat shock, the reporter collapses in protein aggregate centers
Paola Gallardo et al.
Cell reports, 33(6), 108377-108377 (2020-11-12)
Upon acute heat stress (HS), overall mRNA transcription, processing, and export are inhibited, leading to cell growth arrest. However, how cells turn off mRNA metabolism is not fully understood. Here, we show that acute HS results in the segregation and

Questions

1–2 of 2 Questions  
  1. Are these bottles suitable for Avanti J-26 XP centrifuge with a JLA-9.1000 rotor from Beckman Coulter?

    1 answer
    1. Based on the dimensions of this item, the product is not compatible with the JLA-9.1000. This bottle is too wide.

      Helpful?

  2. Are these autoclavable?

    1 answer
    1. This item is autoclavable.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service