Z740499
smartBoat™ disposable paper weighing boats
size M, brown
Synonym(s):
balance accessories, glassine weigh boat, glassine weighing paper, glassine weighing vessel, paper weigh boat, paper weighing vessel, plastic reduction weighing, white paper weigh boat
Select a Size
$93.80
Available to ship on11 April 2025Details
Select a Size
About This Item
$93.80
Available to ship on11 April 2025Details
Recommended Products
material
brown
paper
size
M
packaging
pack of 500 ea
manufacturer/tradename
LevGo Inc 20202
L × W × H
4.5 mm × 4.5 mm × 1.5 mm
color
brown
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- smartBoats will naturally biodegrade in the standard waste stream
- smartBoats control contents like a polystyrene weigh boat and are easier to handle then weigh paper
- Manufactured in an ISO 9001 certified facility in accordance with BGvv, Haccp and FDA regulations
- Made of grease proof paper that meets FDA 21 CFR 176.170, components of paper in contact with aqueous and fatty foods, and with 2I CFR 176.180, contact with dry foods
- Compatible with heat applications with a temperature limitation of 2I0ºC
- Available in a small and medium in both brown for light colored samples and white for dark samples
Sold in packs of 500
Certifications:
USA: ISO 9002 certified facility in accordance with BGvv, Haccp and FDA regulations. FSC Certification
EU: ISO 14001 Certification - Environmental policy, BRC/IOP Certification, ISO 9001 Certification
* Medium brown and white are made in the USA. Small brown and white are made in the European Union.
All products are made from sustainably sourced paper FSC Certified.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service