Skip to Content
Merck
All Photos(2)

Key Documents

Z740307

Disposable Plastic Mini Serological Pipettes

capacity 25 mL

Synonym(s):

cell culture, ergonomic pipetting, pipette, pipetting, pyrogen free, serological, serum, shorty

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

plastic (Disposable)

sterility

sterile; γ-irradiated

feature

graduations Subdivisions 0.5mL
plugged

packaging

pack of 100 ea

manufacturer/tradename

Cole Parmer 2299932

measuring range

accuracy: +1 - 2%

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

Argos brand mini disposable pipettes were developed for tissue culture applications or for use in confined spaces such as laminar hoods. These mini pipettes measure 9 in. and fit most serological pipettors. Smaller size reduces distance between sample and pipettor aiding in more ergonomic pipetting.

Product Details:
  • Polystyrene, gamma sterilized & pyrogen free
  • Easy to read graduations from +1 - 2%
  • Unique filter plug stays put and eliminates lint fibers
  • Color coded rings for easy identification
  • Fiber free paper & plastic wrap ensures quick peel apart opening
  • Dispenser box allows for access of pipettes without opening top; color coded with universal stacking design

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service