Skip to Content
Merck
All Photos(2)

Key Documents

Z740162

Sigma-Aldrich

Phytatray III

L × W × H 9.5 cm × 9.5 cm × 10.5 cm, (recycled polyethylene terephthalate - RPET), sterile; γ-irradiated

Synonym(s):

Phytatray, Phytatray III, Plant Culture Tray

Sign Into View Organizational & Contract Pricing

Select a Size

200 EA
$795.00

$795.00


Check Cart for Availability


Select a Size

Change View
200 EA
$795.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

$795.00


Check Cart for Availability

material

(recycled polyethylene terephthalate - RPET)

sterility

sterile; γ-irradiated

L × W × H

9.5 cm × 9.5 cm × 10.5 cm

application(s)

agriculture

Related Categories

General description

These trays are sterile, disposable plant cell culture vessels made of PETG. They are ideal for numerous plant cell culture applications, such as high throughput screening.

Legal Information

Phytatray is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service