Key Documents
Z740161
LifeSep magnetic separator unit
384F, for 384 well tray
Synonym(s):
cell sorting magnetic seperation unit
Select a Size
1 EA
$2,020.00
$2,020.00
Check Cart for Availability
Select a Size
Change View
1 EA
$2,020.00
About This Item
UNSPSC Code:
41100000
NACRES:
NA.56
$2,020.00
Check Cart for Availability
feature
for 384 well tray
manufacturer/tradename
Dexter Magnetic Technologies 2501007-1
Related Categories
General description
Durable, easy-to-use and designed to accommodate industry standard microtitre plates. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
Features and Benefits
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Optimized field gradient within wells
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What plates is this compatible with?
    1 answer
    1. This product is compatible with most Nunc and Greiner plates.
      Helpful?