Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9864-41

MagRack Maxi

Cytiva 28-9864-41, pack of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.22

packaging

pack of 1 ea

manufacturer/tradename

Cytiva 28-9864-41

availability

not available in Japan

storage temp.

room temp

General description

MagRack Maxi is a magnetic rack for small-scale protein purification and sample enrichment with magnetic beads.

MagRack Maxi can be used for sample volumes of up to 50 ml. Consists of an anodized aluminum housing (blue) with a detachable plastic bar (white) containing a neodymium magnet and this design allows for optimal performance during protein purifications or enrichments. The racks make an excellent complement to the following products (containing magnetic beads) from Cytiva: Protein A Mag Sepharose, Protein A Mag Sepharose Xtra, Protein
G Mag Sepharose, Protein G Mag Sepharose Xtra, His Mag Sepharose Ni, NHS Mag Sepharose, TiO2 Mag Sepharose, and Streptavidin Mag Sepharose.

One of the main advantages of the magnetic beads separation method is the ability to vary the amount of medium as well as the sample volume. MagRack 6 and MagRack Maxi together cover a broad range of sample volumes from low-microliter to high-milliliter purification scales and this further enhances the flexibility of magnetic beads separations.

Application

Highly suitable for use with magnetic beads such as Mag Sepharose platform.

Features and Benefits

  • Provides convenience by making it easy for you to perform small-scale purifications.
  • Allows for optimal performance during protein purifications or enrichments.
  • Enrichment of sample volumes up to 50 mL.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

MagRack is a trademark of Heathrow Scientific
Sepharose is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes immunoprecipitation (immunoaffinity or pull-down techniques).

This page describes manual and automated purification of histidine-tagged recombinant proteins using Cytiva products

Protocols

This page shows magnetic bead-based purification and screening using His Mag Sepharose Ni from Cytiva.

Protein A Mag Sepharose® Xtra and Protein G Mag Sepharose® Xtra enable high-capacity small-scale antibody purification and screening.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service