Skip to Content
Merck
All Photos(1)

Key Documents

Z721107

FAST slides for protein microarrays

1 pad, barcoded, 20 x 51 mm, pk/20

Synonym(s):

microarray slides, protein microarrays

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$1,620.00

$1,620.00


Check Cart for Availability


Select a Size

Change View
20 EA
$1,620.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

$1,620.00


Check Cart for Availability

sterility

non-sterile

packaging

pack of 20

manufacturer/tradename

Maine Mfg 10484182

suitability

suitable for (ELISA format (sandwhich assay) antibody arrays, protein arrays, carbohydrate arrays, reverse phase (micro-Western) arrays using cell or tissue lysates)

Looking for similar products? Visit Product Comparison Guide

General description

Fast Slide 1-Pad
  • Up to 10,000 spots (150 μm spot size, 300 μm pitch)
  • Slide: 1" x 3" (25 x 76 mm)
  • Surface: Nitrocellulose
  • Thickness: 11 μm
FAST Slides are the premier surface for protein microarray applications.

FAST Slides are glass slides coated with a proprietary nitrocellulose polymer. The polymer binds proteins in a noncovalent, irreversible manner and can be probed using the same method as in traditional blots.

The three-dimensional surface of a FAST Slide maintains reactivity of proteins and gives excellent reproducible results. It is usable with fluorescent, chemiluminescent or radiographic detection systems and is compatible with microarray scanners and robots.

Perhaps the most significant advantage of FAST Slides over modified glass surfaces is that the matrix retains arrayed proteins in near-quantitative fashion. This property translates into antibody arrays with unparalleled sensitivity down to antigen concentrations of 1 pg/ml. These qualities make FAST Slides the most reliable surface for microarray experiments and provide a high level of confidence.

Features
  • Superior protein binding capacity
  • Highest sensitivity and dynamic range
  • Excellent long-term stability of printed proteins
  • Compatible with all detection methodologies
  • Compatible with commercially available arraying robots

FAST Slides are suitable for many types of protein microarrays including antibody arrays and microarray Westerns. There are tremendous advantages to using FAST Slides over traditional ELISAs and Westerns including less sample required, better sensitivity, linearity and quantitation. The major advantage is that hundreds or thousands of antibodies or samples can be screened simultaneously.

Compatible detection methodologies

FAST Slides are compatible with fluorescent, chemiluminescent or radiographic detection systems.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service