Skip to Content
Merck
All Photos(1)

Key Documents

GE27-4575-01

DNA (Calf Thymus), activated

Cytiva 27-4575-01, pack of 1 ea

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$568.00

$568.00


Check Cart for Availability


Select a Size

Change View
500 G
$568.00

About This Item

UNSPSC Code:
41116006
NACRES:
NA.52

$568.00


Check Cart for Availability

packaging

pack of 1 ea

manufacturer/tradename

Cytiva 27-4575-01

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

DNA (Calf Thymus)

  • Nonmethylated DNA (at a concentration of 500 μg/ml) for preparation of molecular weight markers.
  • Methylated DNA and nonmethylated DNA (at a concentration of 500 μg/ml) for determining methylation sensitivity of restriction enzymes.
  • High-quality DNAs from viral and eukaryotic sources used for preparation of assay reagents.
  • Activated calf thymus DNA at 25 A260 μ/ml (treated briefly with DNase I)—for preparing substrate used in the study of DNA polymerases.

Features and Benefits

  • High-quality DNAs from viral and eukaryotic sources used for preparation of assay reagents.
  • Nonmethylated DNA (at a concentration of 500 μg/mL) for preparation of molecular weight markers.
  • Methylated DNA and nonmethylated DNA (at a concentration of 500 μg/mL) for determining methylation sensitivity of restriction enzymes.
  • Activated calf thymus DNA at 25 A260 μ/mL (treated briefly with DNase I)—for preparing substrate used in the study of DNA polymerases.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service