Key Documents
Z694665
3D Biotek Polycaprolactone (PCL) scaffold inserts
for 3D bioreactor 12 well chamber, fiber diam. ~300 μm, ~ 300 μm spacing
Select a Size
10 EA
$374.00
$374.00
Estimated to ship on14 April 2025
Select a Size
Change View
10 EA
$374.00
About This Item
UNSPSC Code:
41122107
NACRES:
NB.22
$374.00
Estimated to ship on14 April 2025
material
(polycaprolactone (PCL))
sterility
sterile
feature
lid: no
~ 300 μm spacing
manufacturer/tradename
3D Biotek PCL303012-BR
fiber diam.
~300 μm
working volume
~300 μL
pore size
300 μm (Fiber spacing)
binding type
non-treated surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)