Z688797
Nunc® UpCell™ Surface cell culture multidish
size 12 wells, case of 6 ea
Synonym(s):
cell culture multidish, tissue culture multidish
Select a Size
$560.00
Select a Size
About This Item
$560.00
Recommended Products
material
clear plate
clear wells
sterility
sterile
sterile
feature
lid
membrane not included
w/ lid and airvent
packaging
case of 6 ea
manufacturer/tradename
Nunc 174900
dish L × W
128 mm × 86 mm
external L × W
128 mm × 86 mm
size
12 wells
surface area
3.5 cm2 , culture area (cm2)/well
working volume
2 mL
binding type
(UpCell Surface)
Related Categories
General description
- Harvesting of cells with intact surface proteins
- No trypsinization - preserve cell surface proteins
- No physical force - get high cell viability
- Releases adherent cells by reduction of temperature of the cell culture
- For culture passaging, single-cell analyses and cell transplantation research
- Enables harvesting of cell sheets and creations of 3D tissue models held together by normal cell junctions and extracellular matrix
- Minimal hands-on time
- For research and single-use only
- It is quick, clean and simple-just reduce the temperature
Note: Suitable for the harvesting of cells with intact surface proteins, for cell culture passage, single-cell analyses and cell transplantation research.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)