Yes, Z569771 is the same as A2473, Thermo MBP Molecular BioProducts 2140, and Krackeler 545-2140-05. Customers may need to confirm the pack sizes and tray numbers, but these catalog numbers should all be the same tip.
Z569771
ART® self-sealing barrier pipette tips
ART 10 Reach, volume range 0.1-10 μL, sterile
Synonym(s):
pipet tips, pipette tips
Select a Size
$429.00
Select a Size
About This Item
RNase free
barrier
$429.00
Recommended Products
material
colorless
sterility
sterile
feature
DNase free
RNase free
barrier
packaging
pack of 960 ea
manufacturer/tradename
Molecular BioProducts 2140
technique(s)
PCR: suitable
maximum volume
10 μL
volume range
0.1-10 μL
volume
0.1—10 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Gel loading: for convenient loading of acrylamide or agarose gels.
- Wide orifice genomic: for handling genomic DNA, fragile cell lines, and other viscous materials.
- Solvent safe: withstands acids, bases, and aggressive organic solvents.
- Genetic studies
- Forensics
- General PCR work
- Radioisotope sampling
Features and Benefits
- Free of DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
- Available in sterile and non-sterile options to suit different needs.
- Offered in various packaging options for convenience and flexibility.
- Manufactured in adherence to the highest quality standards set by the industry.
Packaging
4800EA = 5 packs of 960 tips (50 trays of 96 tips)
800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Is the Z569771 the same as the A2473? I want A2473 in the photo on the webpage for Z569771, but the picture is confusing with all the different styles of tips.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service