Skip to Content
Merck
All Photos(1)

Key Documents

Z717207

ART® self-sealing barrier pipette tips

ART 200G, maximum volume 200 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

960 EA
$405.00
4800 EA
$1,510.00

$405.00


Check Cart for Availability


Select a Size

Change View
960 EA
$405.00
4800 EA
$1,510.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
200 μL
sterility:
sterile
feature:
barrier
packaging:
pack of 960 ea (racked)
manufacturer/tradename:
Molecular BioProducts 2069G

$405.00


Check Cart for Availability

material

colorless

sterility

sterile

feature

barrier

packaging

pack of 960 ea (racked)

manufacturer/tradename

Molecular BioProducts 2069G

technique(s)

PCR: suitable

maximum volume

200 μL

volume

200 μL

compatibility

for use with

  • Biohit Proline-Electronic 10-100 μl
  • Biohit Proline-Variable 5-50 μl/50-200 μl
  • Eppendorf Research/Reference 20-200 μl
  • Eppendorf Titerman 4908 30-300 μl
  • Finnpipette 4027/4500 Digital 40-200 μl
  • Finnpipette Multi-Channel (Limited Draw) 50-300 μl
  • Mettler-Toledo VoluMate 20-200 μl
  • Nichiryo BenchMate 5-100 μl/20-200 μl
  • Pipetman P-200 20-200 μl
  • Rainin EDP 2 20-250 μl
  • Rainin EDP Plus 20-250 μl
  • Rainin Pipet Plus 20-200 μl
  • Rainin Pipetman 20-200 μl

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

A Genomic tip refers to a tip with a wide-bore orifice. This kind of tip is perfect for pipetting more viscous samples such as blood as well as for preventing the shearing of genomic DNA during sample processing and preparation.

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service