Skip to Content
Merck
All Photos(2)

Key Documents

Z378674

Sigma-Aldrich

Sigma-Solohill microcarrier beads

Collagen coated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

polystyrene beads

bead size

125-212 μm

particle size

125-212 μm

density

1.02 g/cm3 (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

The porcine collagen surface layer promotes attachment and monolayer growth.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service