Skip to Content
Merck
All Photos(2)

Key Documents

Z290416

Sigma-Aldrich

Sigma-Aldrich® vacuum filtration assembly

for 47 mm filters with glass support, NS 40/35 joints, 4 L flask (included)

Synonym(s):

Filtration Assembly, Vacuum Filtration Setup

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,290.00

$1,290.00


Available to ship on22 April 2025Details



Select a Size

Change View
1 EA
$1,290.00

About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

$1,290.00


Available to ship on22 April 2025Details


funnel volume

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Same as Z290432 but with NS 40/35 jont (male on flask, female on funnel support) instead of rubber stopper.
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.

Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).

For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service