Skip to Content
Merck
All Photos(3)

Key Documents

WH0004684M1

Sigma-Aldrich

Monoclonal Anti-NCAM1 antibody produced in mouse

clone 3G12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD56, Anti-MSK39, Anti-NCAM, Anti-neural cell adhesion molecule 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$484.00

$484.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1116


Select a Size

Change View
100 μG
$484.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$484.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1116

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCAM1(4684)

General description

Neural cell adhesion molecule (NCAM/CD56) is a calcium-independent binding protein. It is located on human chromosome 11q23.1. This molecule belongs to the immunoglobulin superfamily. NCAM1 is present in neurons and glial cells.

Immunogen

NCAM1 (AAH47244, 611 a.a. ~ 710 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
EPSAPKLEGQMGEDGNSIKVNLIKQDDGGSPIRHYLVRYRALSSEWKPEIRLPSGSDHVMLKSLDWNAEYEVYVVAENQQGKSKAAHFVFRTSAQPTAIP

Biochem/physiol Actions

Neural cell adhesion molecule (NCAM/CD56) participates in homophilic cell–cell and heterophilic cell–matrix interactions. It controls the movement of cells and condensation during skeletal development. This protein participates in synaptic plasticity, neurodevelopment and neurogenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of neural cell adhesion molecule and polysialic acid in human bone marrow-derived mesenchymal stromal cells
Skog MS, et al.
Stem Cell Research & Therapy, 7(1), 113-113 (2016)
NCAM1 association study of bipolar disorder and schizophrenia: polymorphisms and alternatively spliced isoforms lead to similarities and differences
Atz ME, et al.
Psychiatric Genetics, 17(2), 55-67 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service