Skip to Content
Merck
All Photos(4)

Key Documents

WH0000632M1

Sigma-Aldrich

Monoclonal Anti-BGLAP antibody produced in mouse

clone 2D4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BGP, Anti-bone gamma-carboxyglutamate (gla) protein (osteocalcin)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$548.00

$548.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1704


Select a Size

Change View
100 μG
$548.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$548.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1704

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1λ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BGLAP(632)

Immunogen

BGLAP (NP_954642, 52 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YLYQWLGAPVPYPDPLEPRREVCELNPDCDELADHIGFQEAYRRFYGPV

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniella Da Silva et al.
Regenerative therapy, 24, 602-616 (2023-11-30)
Worldwide, osteoporosis is the utmost predominant degenerative bone condition. Stem cell regenerative therapy using adipose-derived mesenchymal stem cells (ADMSCs) is a promising therapeutic route for osteoporosis. Photobiomodulation (PBM) has sparked considerable international appeal due to its' ability to augment stem
Qinglin Han et al.
Molecular medicine reports, 16(5), 7591-7596 (2017-09-26)
The aim of the present study was to investigate the effects of cobalt nanoparticles (CoNPs) on the proliferation and differentiation of human osteoblasts in vitro, and to investigate the molecular mechanisms via which CoNPs affect proliferation and differentiation of osteoblasts. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service