Skip to Content
Merck
All Photos(1)

Key Documents

U5754

Sigma-Aldrich

Urodilatin human

≥97% (HPLC)

Synonym(s):

Thr-Ala-Pro-Arg-Atrial Natriuretic Peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C145H234N52O44S3
CAS Number:
Molecular Weight:
3505.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

biological source

human

Quality Level

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ~70%

technique(s)

activity assay: suitable

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCSC)NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)CNC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC(=O)CNC1=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc4ccc(O)cc4)C(O)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CO)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]5CCCN5C(=O)[C@H](C)NC(=O)[C@@H](N)[C@@H](C)O

InChI

1S/C145H234N52O44S3/c1-11-72(6)112-137(238)171-60-106(208)172-73(7)113(214)177-86(40-41-103(146)205)122(223)190-95(63-198)116(217)170-61-108(210)175-88(51-70(2)3)114(215)169-62-109(211)176-100(133(234)187-92(56-104(147)206)127(228)193-97(65-200)130(231)186-91(54-77-27-16-13-17-28-77)126(227)180-82(31-20-45-163-142(153)154)119(220)189-94(139(240)241)55-78-36-38-79(204)39-37-78)68-243-244-69-101(134(235)185-90(53-76-25-14-12-15-26-76)115(216)168-58-105(207)167-59-107(209)174-80(29-18-43-161-140(149)150)117(218)182-87(42-50-242-10)123(224)188-93(57-110(212)213)128(229)181-85(124(225)196-112)34-23-48-166-145(159)160)195-132(233)99(67-202)194-131(232)98(66-201)192-120(221)83(32-21-46-164-143(155)156)178-118(219)81(30-19-44-162-141(151)152)179-125(226)89(52-71(4)5)184-129(230)96(64-199)191-121(222)84(33-22-47-165-144(157)158)183-135(236)102-35-24-49-197(102)138(239)74(8)173-136(237)111(148)75(9)203/h12-17,25-28,36-39,70-75,80-102,111-112,198-204H,11,18-24,29-35,40-69,148H2,1-10H3,(H2,146,205)(H2,147,206)(H,167,207)(H,168,216)(H,169,215)(H,170,217)(H,171,238)(H,172,208)(H,173,237)(H,174,209)(H,175,210)(H,176,211)(H,177,214)(H,178,219)(H,179,226)(H,180,227)(H,181,229)(H,182,218)(H,183,236)(H,184,230)(H,185,235)(H,186,231)(H,187,234)(H,188,224)(H,189,220)(H,190,223)(H,191,222)(H,192,221)(H,193,228)(H,194,232)(H,195,233)(H,196,225)(H,212,213)(H,240,241)(H4,149,150,161)(H4,151,152,162)(H4,153,154,163)(H4,155,156,164)(H4,157,158,165)(H4,159,160,166)/t72-,73-,74-,75+,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,111-,112-/m0/s1

InChI key

IUCCYQIEZNQWRS-DWWHXVEHSA-N

Gene Information

human ... NPPA(4878)

Amino Acid Sequence

Thr-Ala-Pro-Arg-Ser-Leu-Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Met-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr [Disulfide Bridge: 11-27]

Application

Human Urodilatin can be used as a test compound for examining the activity and mechanisms of action of urodilatin in bovine bronchi. It used to investigate the effect on bronchodilatation and protect against subsequent challenge with alpha-human atrial natriuretic peptide (ANP).

Biochem/physiol Actions

Urodilatin is a natriuretic factor belongs to atrial natriuretic peptide family which along with four cardiac hormones decreases human renal carcinoma cell numbers. Urodilatin induces cGMP synthesis as well as stimulates cGMP accumulation by threefold in inner medullary collecting ducts (IMCD).
Urodilatin is a natriuretic factor that is produced in the kidney and secreted into the urine. It regulates water and sodium reabsorption in the collecting duct.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Bailly
Kidney international. Supplement, 65, S29-S35 (1998-04-29)
The thick ascending limb of Henle's loop (TAL) is involved in the urinary dilution/concentration process by actively reabsorbing NaCl through a complex mechanism. Some years ago, compelling evidence was provided that cAMP stimulates NaCl reabsorption through the activation of adenylyl
M Heringlake et al.
The American journal of physiology, 277(3 Pt 2), F347-F351 (1999-09-14)
The findings about mechanisms regulating production and excretion of urodilatin [ANP-(95-126)], a member of the atrial natriuretic peptide (ANP) family, are controversial. To elucidate a possible relationship between arterial blood pressure and renal urodilatin excretion, we studied the effects of
W G Forssmann et al.
Histochemistry and cell biology, 110(4), 335-357 (1998-10-29)
This review focuses on some selected aspects of the endocrine heart and natriuretic peptides. The endocrine heart is composed of specific myoendocrine cells of the cardiac atria. The myoendocrine cells synthesize and secrete the natriuretic peptide hormones which exhibit natriuretic
J E Nally et al.
British journal of pharmacology, 114(7), 1391-1396 (1995-04-01)
1. This study examined the activity and mechanisms of action of urodilatin in bovine bronchi. For comparison, the ability of urodilatin to evoke bronchodilatation or protect against subsequent challenge was compared to that of the closely related peptide alpha-human atrial
J Koike et al.
Journal of the American Society of Nephrology : JASN, 3(10), 1705-1709 (1993-04-01)
Urodilatin is found in the urine and is thought to be produced in the kidney. The effect of urodilatin on cGMP accumulation in the kidney was investigated. cGMP accumulation by urodilatin and atrial natriuretic peptide (ANP) were compared with tubule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service